Learn More
Medchemexpress LLC Setipiprant | 866460-33-5 | 98.2% | 402.42 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY16635
Encompass_Preferred
Setipiprant is an orally active and selective CRTH2 antagonist. It interacts with the hCRTH2 receptor with an IC50 value of 6 nM and inhibits prostanoid receptors hDP1 and hEP2. This compound is suitable for research related to asthma and rhinitis.
  • Orally active and selective CRTH2 antagonist
  • Interacts with hCRTH2 receptor with IC50 of 6 nM
  • Inhibits prostanoid receptors hDP1 and hEP2
  • Used for research related to asthma and rhinitis
Catalog No. 50-003-95411