Learn More
Medchemexpress LLC Setmelanotide | 920014-72-8 | 99.9% | 10 MG

Supplier: Medchemexpress LLC HY1987010MG
Setmelanotide is a research-grade peptide agonist selective for the melanocortin 4 receptor (MC4R). It exhibits sub-nanomolar potency (EC50 ≈ 0.27 nM human, 0.28 nM rat), has a molecular weight of 1117.31, and is supplied as a high-purity white to off-white solid. The peptide is highly soluble in water and requires cold, sealed storage for long-term stability.
- Selective melanocortin 4 receptor (MC4R) agonist with sub-nanomolar potency.
- High chemical purity (99.93%).
- White to off-white solid physical form.
- Water solubility approximately 62.5 mg/mL (requires ultrasonic).
- Stable as powder at -80°C for up to 2 years and at -20°C for 1 year.
- Contains a defined peptide sequence with a disulfide bridge.
- Suitable for pharmacology and receptor biology research applications.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.