Learn More
TARGETMOL CHEMICALS INC SETROBUVIR 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T287625MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Setrobuvir (ANA-598) is an orally active non-nucleoside HCV NS5B polymerase inhibitor with inhibitory effects on de novo RNA synthesis and primer extension with IC50s between 4 and 5 nM. setrobuvir shows good binding affinity for SARS-CoV-2 RdRp and induces inhibition of RdRp. purity: 98%
Catalog No. 50-258-0494