Learn More
Cole-Parmer Raven Environmental C-10102 Settlometer Kit, acrylic, 1.4 L, dual scale % solids and SSV
Supplier:  Cole-Parmer 0547600
Encompass
This clear plastic settlometer includes mixing paddle, snap-on lid, and instructions. Order the optional interval timer separately below to measure sludge settling rates.
Catalog No. 50-632-416