Learn More
Hach Company Settlometer Kit
Supplier:  Hach Company 2115700
Encompass
Complete kit for analyzing activated sludge process Includes a transparent polycarbonate 2-L jar with cover Jar has easy-to-read white graduations and is impact-resistant Custom polypropylene paddle allows agitation of sample without scratching the jar Also includes 10 polycarbonate centrifuge tubes (15 mL graduated) All components chemically resistant at pH range 3-10
Catalog No. 50-002-57089