Learn More
Medchemexpress LLC Sevabertinib | 2521285-05-0 | 99.9% | 484.93 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY164718
Encompass_Preferred
Sevabertinib (BAY 2927088) is an orally active and reversible dual EGFR-HER2 inhibitor with IC50 values of <0.5 nM for wild-type HER2, HER2 A775insYVMA, wild-type EGFR, and EGFR D770_N771insSVD. It demonstrates anticancer activity against lung cancer.
  • Orally active and reversible dual EGFR-HER2 inhibitor.
  • Potent inhibition of multiple HER2 and EGFR forms (IC50 <0.5 nM).
  • Shows anticancer activity against lung cancer.
  • Inhibits proliferation of isogenic Ba/F3 cells with HER2 exon 20 insertions or resistance mutations.
  • Potently inhibits proliferation of NCI-H1781 and NCI-H2170 lung cancer cell lines.
  • Demonstrates tumor growth inhibition in PDX mouse models of NSCLC.
  • Shows antitumor efficacy in NCI-H1781 xenograft mouse model.
Catalog No. 50-003-94787