Learn More
Medchemexpress LLC Sevasemten (EDG-5506) | 2417395-15-2 | 99.3% | 381.28 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1487995MG
Encompass_Preferred
Sevasemten is an orally active, selective allosteric inhibitor of skeletal muscle myosin. It protects skeletal muscle from contraction-induced injury, reduces muscle damage biomarkers and fibrosis, and increases muscle strength and activity in Duchenne muscular dystrophy disease models.
  • Orally active, selective allosteric inhibitor of skeletal muscle myosin
  • Protects skeletal muscle from contraction-induced injury
  • Decreases muscle damage biomarkers and fibrosis
  • Increases muscle strength and activity in Duchenne muscular dystrophy disease models
  • Reduces the force of fast skeletal muscle fibers
  • Inhibits fast myofibril ATPase
  • Protects muscles from contraction damage
  • Ameliorates Duchenne muscular dystrophy in animal models
Catalog No. 50-003-61577