Learn More
Selleck Chemical LLC Sevelamer Carbonate S4134-50mg
SDP
Supplier:  Selleck Chemical LLC S413450MG
Encompass_Preferred
Sevelamer carbonate is a non-absorbed phosphate binding crosslinked polymer with the same polymeric structure as sevelamer hydrochloride in which carbonate replaces chloride as the counterion
Catalog No. 50-310-9948