Learn More
Abcam SEW 2871, S1P1 agonist, 10MG
Supplier:  Abcam AB12098310MG
Encompass_Preferred
MW 440.4 Da, Purity >98%. Potent, selective sphingosine-1-phosphate (S1P1) agonist (EC50 = 13 nM). Does not activate S1P2, S1P3, S1P4 or S1P5 receptors. Cell-permeable and active *in vivo*.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7062