Learn More
Lonza Walkersville SF Cell Line 384-well Nucleofector Kit
Supplier:  Lonza Walkersville V5SC2010
Encompass
The SF Cell Line 384-well Nucleofector Kit is one of our three kits suited for high-throughput transfection of cell lines when working with the 384-well HT Nucleofector System.Benefits:High transfection efficiencies in difficult-to-transfect cell linesCells maintain functionality post transfectionSeamless integration into automated liquid handling environmentsConditions are transferable between 4D-Nucleofector System, 96-well Shuttle System and 384-well Nucleofector System Applications:Transfection of low cell numbers down to 2 x 10⁴ cell linesHigh-throughput applications, e.g. screens of arrayed cDNA, RNAi or CRISPR libraries Content:90 mL SF Cell Line Nucleofector Solution20 mL Supplement 150 µg pmaxGFP Vector (1 μg/μl in 10 mM Tris pH 8.0)10 x 384-well Nucleocuvette Plate (20 μL)
Catalog No. NC1999827
Add to Cart
Request a Quote