Learn More
Selleck Chemical LLC SF1670 5mg
SDP
Supplier:  Selleck Chemical LLC S73105MG
Encompass_Preferred
SF1670 is a highly potent and specific PTEN inhibitor with IC50 of 2 ?M. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3265