Learn More
Abcam SF1670, PTEN inhibitor, 5MG
Supplier:  Abcam AB1413035MG
Encompass_Preferred
MW 307.3 Da, Purity >98%. Selective PTEN inhibitor (IC₅₀ = 2 μM). Elevates intracellular PIP3 signaling. Reduces neutrophil functions, including phagocytosis, oxidative burst, polarization, and chemotaxis. Active in vivo.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6701