Learn More
Medchemexpress LLC SF20/MYDGF Mouse 2ug
SDP
Supplier:  Medchemexpress LLC HYP733032UG
Encompass_Preferred
SF20/MYDGF protein promotes cardiac myocyte survival and adaptive angiogenesis after myocardial infarction (MI) It stimulates endothelial cell proliferation via MAPK1/3 STAT3 and CCND1 signaling and inhibits cardiac myocyte apoptosis through PI3K/AKT signaling Given its ability to enhance cardiac function and angiogenesis SF20/MYDGF protein could be a valuable therapeutic target for improving outcomes in MI patients SF20/MYDGF Protein Mouse is the recombinant mouse-derived SF20/MYDGF expressed by E coli with tag-free
Catalog No. 50-001-80431