Learn More
Expression Systems Sf21 cells, Suspension in ESF 921, approx. 1M cells/mL (50mL/125mL flask)
SDP
Supplier:  Expression Systems 94003S
Encompass_Preferred
Expression Systems offers insect cell lines adapted to serum-free suspension culture in either ESF 921 or ESF AF. The availability of pre-adapted cells saves significant time and expense associated with the adaptation of cultures.
The Sf21 insect cell line (IPLB-Sf-21-AE) is derived from ovarian cells of Spodoptera frugiperda. Sf21 cells are adapted for suspension culture in ESF 921 or ESF AF and are available as a frozen vial or suspension culture.
• Recombinant protein expression from a variety of expression platforms
• Good growth in adherent or suspension culture
• Quality and performance testing
To download Instructions for Use and/or COA please visit: www.expressionsystems.com
Catalog No. NC0681197