Learn More
Medchemexpress LLC Sf2523 | 1174428-47-7 | 99.3% | 371.41 g/mol | C19H17NO5S | 1 ML
SDP
Supplier:  Medchemexpress LLC HY10114610MM 1ML
Encompass_Preferred
SF2523 is a small-molecule kinase and bromodomain inhibitor with potent activity against PI3K isoforms, DNA-PK, BRD4 (BD1), and mTOR. Supplied as a solid or as a 10 mM solution in DMSO (1 mL), it is intended for biochemical and cellular research into PI3K/Akt/mTOR signaling, DNA damage response, and epigenetic regulation. The compound is high purity and has documented storage recommendations for powder and solution formats.
  • Potent inhibition of DNA-PK and PI3K with low-nanomolar IC50 values.
  • Activity against BRD4 and mTOR enables dual pathway modulation.
  • Available as solid or 10 mM DMSO solution for assay flexibility.
  • High purity (>99%) suitable for research applications.
  • Datasheet and safety data sheet available for handling and storage guidance.
Catalog No. 50-002-13374