Learn More
Medchemexpress LLC SF2523 | 1174428-47-7 | MFCD31382129 | 99.3% | 371.41 | C19H17NO5S | 25 MG
SDP
Supplier:  Medchemexpress LLC HY10114625MG
Encompass_Preferred
SF2523 is a small-molecule research inhibitor that targets PI3K isoforms and related kinases, exhibiting low-nanomolar activity against DNA-dependent protein kinase and sub-100 nM activity against PI3Kα. Supplied as a high-purity solid for in vitro studies, the product includes manufacturer guidance for solubility and storage.
Catalog No. 50-002-11230