Learn More
ABclonal Technology SF3A3 Rabbit mAb
SDP
Supplier:  ABclonal Technology A2092720UL
Encompass_Preferred
This gene encodes subunit 3 of the splicing factor 3a protein complex The splicing factor 3a heterotrimer includes subunits 1 2 and 3 and is necessary for the in vitro conversion of 15S U2 snRNP into an active 17S particle that performs pre-mRNA splicing Subunit 3 interacts with subunit 1 through its amino-terminus while the zinc finger domain of subunit 3 plays a role in its binding to the 15S U2 snRNP This gene has a pseudogene on chromosome 20 Alternative splicing results in multiple transcript variants
Catalog No. 50-000-08155