Learn More
Aviva Systems Biology Corp SF3B1 Antibody - N-terminal region (ARP40733_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40733_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against SF3B1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SF3B1. Alternate names: MDS, PRP10, Hsh155, PRPF10, SAP155, SF3b155ApplicationWB, IHC. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rat, Zebrafish
Catalog No. 50-014-57603