Learn More
Cygnus Technologies Cygnus Technologies Sf9 HCP ELISA Kit, 3rd Generation, High Sensitivity 0.3 ng/mL, For Research and Manufacturing Use Only, Kit
Supplier:  Cygnus Technologies F1040
Encompass
The Cygnus Technologies Sf9 HCP ELISA Kit, 3G provides a sensitive and objective assay for detecting host cell protein impurities in biopharmaceutical products produced with Sf9 insect cells. It leverages high-affinity goat polyclonal antibodies, ensuring broad reactivity critical for process control and product release testing, thus enhancing product purity and safety. Unlike older versions, this 3rd generation kit offers an improved quantitation range and sensitivity, making it essential for quality control in bioprocessing.
Catalog No. NC2598349