Learn More
ATCC Sf9; Ovary; Fall Armyworm; (Spodoptera frugiperda)
Supplier:  ATCC CRL1711
Encompass
Sf9 is a cell line exhibiting epithelial morphology that was derived from pupa ovarian tissue of a fall armyworm. This cell line can be used to replicate baculovirus expression vectors.
Catalog No. 50-238-2535
 Restricted Item