Learn More
ATCC Sf9-ET; Ovary; Fall Armyworm; (Spodoptera frugiperda)
Supplier:  ATCC CRL3357
Encompass
Sf9-ET is a cell line with rounded morphology that was isolated in 1993 from the ovary of a fall armyworm (Spodoptera frugiperda). The Sf-9 cell line has been modified to express eGFP upon infection with baculovirus. It was deposited by the Frederick National Laboratory for Cancer Research.
Catalog No. 50-238-2537
 Restricted Item