Learn More
Biotage Sfär DLV Column with 25g of Silicia
SDP
Supplier:  Biotage DLV04450050
Encompass
Biotage DLV (Dry Load Vessel) systems provide a convenient in-line means for introducing samples. Users adsorb liquid samples onto the 25 grams of pre-packed adsorbent inside the DLV and then attach to any Biotage Sfar column or flash chromatography column with luer fittings. Biotage DLVs work well on Biotage Sfar Duo columns when larger quantities of sorbant material are needed.Come in a pack of 20. These vessels come pre-packed with 25 grams of silica gel and have approximately 25 mL of headspace to accomdate liquid loading.Compatible with Biotage DLV plunger, part number DLV-050.
Catalog No. NC1606651