Learn More
Biotage Sfär Silica Samplet 200 to 400g Column
SDP
Supplier:  Biotage SAS04450350
Encompass
Creating dried, adsorbed samples will improve the separation performance on any cartridge. Biotage Samplet are designed for quick and convenient loading of concentrated samples.After the sample has been applied to the Samplet, the solvent is allowed to evaporate. The Samplet is then inserted into the Biotage Sfär Duo columns and you are ready to go.These Samplets are compatible with 200 to 400 gram columns and contain approximately 30 grams of silica sorbent material. Shipped in packs of 6.
Catalog No. NC1607527