Learn More
Biotage Sfär Silica HC Duo 20µm 50g - High Capacity Column
SDP
Supplier:  Biotage FSUD04430050
Encompass
Biotage Sfär Silica High Capacity Duo 20 μm is the choice when you need to run the best possible separation with the smallest column and greenest amount of solvent. You also have the flexibility to load your sample in six different ways.This 50g high capacity column has the separation capacity of a 100g standard capacity column.Sfär means “Sphere” in the Swedish mother tongue of Biotage, and is the name for our columns as they contain spherical silica with higher surface area compared to irregular silica. Which means you use a smaller column with tight elution bands and pure fractions.Sfär columns are quality tested for separation efficiency and peak symmetry. Each CE-marked column is created using inert, food grade plastics for lower extractables, cleaner fractions, and packed to provide excellent resolution and high pressure tolerance with a rating of 17 bar or 247 psi.These 50g columns come in convenient dispenser boxes containing 10 columns
Catalog No. NC1606536