Learn More
Bachem TRAP-5 amide Trifluoroacetate
SDP
Supplier:  Bachem 4031273.0100
Encompass_Preferred
TRAP-5 amide Trifluoroacetate, H-Ser-Phe-Leu-Leu-Arg-NH2, 141923-41-3, C30H51N9O6, Mr 633.79, 100mg. SFLLRamide, Thrombin Receptor Activator Peptide 5 amide, PAR-1 (1-5) amide (human), Proteinase Activated Receptor 1 (1-5) amide (human), Thrombin Receptor (1-5) amide (human), Coagulation Factor II Receptor (1-5) amide (human). SFLLR-amide, thrombin receptor agonist peptide.
Catalog No. 50-194-6789