Learn More
Medchemexpress LLC SFRP2 Human HEK293 50ug | 50ug
SDP
Supplier:  Medchemexpress LLC HYP7783550UG
Encompass_Preferred
SFRP2 Protein is a soluble crimson-associated protein that promotes tumor development by activating the Wnt/ -catenin signaling pathway The methylation of SFRP2 Protein DNA is associated with tumor development SFRP2 Protein Human (HEK293 His solution) is the recombinant human-derived SFRP2 protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-87688