Learn More
Aviva Systems Biology Corp SFTPA2 Antibody - N-terminal region (ARP64034_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64034_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SFTPA2. It was validated on Western Blot. . Alternate names: COLEC5, FLJ50594, FLJ50597, FLJ51953, FLJ79091, FLJ93678, MGC133169, MGC133366, MGC189714, MGC189761, PSAP, SFTP1, SFTPA2B, SPA2, SPAIIApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-288-150