Learn More
Andwin Scientific SFX PRTBLE HNDHLD FBR LSER 50W
SDP
Supplier:  Andwin Scientific SFX7070
Encompass_Preferred
SFX PORTABLE HANDHELD FIBER LASER ENGRAVER 50W WITH RECHARGEABLE BATTERY LIGHT WEIGHT AND EASY TO MOVER LASER WITH METAL AND NON-METAL MARKINGS 70*70MM
Catalog No. NC2884275
Only null left
Add to Cart
Request a Quote