Learn More
Molecular Dimensions SG2 1ML HT BLOCK 96X1ML

Supplier: Molecular Dimensions MD1132
- High-throughput crystallization block design.
- 1 ml well capacity.
- 96 wells available.
- Format caters to 96 x 1 ml configurations.
- Application focused on protein crystallization screening.
- ShotGun screening method utilized.
- Features 96 successful conditions identified from the Protein Data Bank.
- Facilitates easy optimization of crystallization conditions.
- Reduces occurrence of co-crystallization hits.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.