Learn More
Medchemexpress LLC HY-101161 1mg , SG3199 CAS:1595275-71-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1011611MG
Encompass_Preferred
HY-101161 1mg SG3199 CAS: 1595275-71-0 SG3199 is a cytotoxic DNA minor groove interstrand crosslinking pyrrolobenzodiazepine (PBD) dimer. SG3199 is the released warhead component of the ADC payload Tesirine (SG3249). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9286