Learn More
Medchemexpress LLC SG3199-Val-Ala-PAB | 1595275-61-8 | MFCD34469129 | 96.9% | 922.03 | C49H59N7O11 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY101152100MG
Encompass_Preferred
SG3199-Val-Ala-PAB is a synthetic intermediate for the preparation of Tesirine, an agent-linker conjugate employed in antibody-drug conjugate (ADC) research. Supplied as a white to off-white solid, it is provided for research use and requires protected, low-temperature storage to maintain stability.
  • Used as an intermediate in ADC linker synthesis.
  • Contains a Val-Ala protease-cleavable dipeptide and a PAB spacer.
  • High purity suitable for research applications.
  • Solid form stored at -20°C; protect from light and oxygen.
  • High solubility in DMSO; ultrasonic agitation recommended for dissolution.
Catalog No. 50-002-11658
Add to Cart
Request a Quote