Learn More
Medchemexpress LLC Sg3199-val-ala-pab | 1595275-61-8 | MFCD34469129 | 96.9% | 922.03 g/mol | C49H59N7O11 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY10115210MG
Encompass_Preferred
SG3199-Val-Ala-PAB is a synthetic intermediate used in the assembly of Tesirine-based antibody-drug conjugates (ADCs). It is intended for research use only and is provided with supporting documentation for safe handling and experimental traceability. The compound features a valine-alanine-para-aminobenzyl (Val-Ala-PAB) cleavable linkage, formula C49H59N7O11, molecular weight ~922 g/mol, and CAS 1595275-61-8.
  • High purity suitable for synthetic applications and analytical characterization.
  • Provides a cleavable valine-alanine-PAB linkage for controlled payload release.
  • Packaged in small-mass quantities for medicinal chemistry and ADC development workflows.
  • Accompanied by certificate of analysis and safety data sheet for traceability.
  • Intended for research use only; not for human or diagnostic use.
Catalog No. 50-002-11659