Learn More
Selleck Chemical LLC SGC 0946 10mg
SDP
Supplier:  Selleck Chemical LLC S707910MG
Encompass_Preferred
SGC 0946 is a highly potent and selective DOT1L methyltransferase inhibitor with IC50 of 0.3 nM in a cell-free assay, is inactive against a panel of 12 PMTs and DNMT1. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3252