Learn More
Apexbio Technology LLC SGI-1776 free base, 10mg. Cas: 1025065-69-3 MFCD: MFCD16659064. Pim kinase inhibitor,ATP-competitive
SDP
Supplier:  Apexbio Technology LLC A419210
Encompass_Preferred
SGI-1776 free base, N-((1-methylpiperidin-4-yl)methyl)-3-(3-(trifluoromethoxy)phenyl)imidazo[1,2-b]pyridazin-6-amine, is a potent ATP-competitive inhibitor of the serine/threonine family of Pim kinase, an enzyme regulating cell survival. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4903