Learn More
Medchemexpress LLC VHL ligand 8 | 2701565-76-4 | MFCD01735918 | 98.0% | 443.49 g/mol | C23H29N3O6 | 5 MG

Supplier: Medchemexpress LLC HY1330455MG
VHL Ligand 8 is a small-molecule ligand for the von Hippel-Lindau (VHL) E3 ligase used as a building block in PROTAC synthesis and targeted protein degradation studies. It is provided as a high-purity solid (C23H29N3O6, 443.49 g/mol) and includes documented solubility and storage recommendations for in vitro and in vivo formulation.
- High purity (98.0%).
- Well-characterized identity: CAS 2701565-76-4; formula C23H29N3O6.
- Solid form suitable for storage and handling.
- Compatible with common solvents; soluble in DMSO and formable for in vivo use.
- Stable under recommended storage conditions: powder stored at -20°C or 4°C.
- Suitable for PROTAC synthesis and cellular degradation assays.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.