Learn More
Selleck Chemical LLC SGI-1776 free base 50mg 1025065-69-3
SDP
Supplier:  Selleck Chemical LLC S219850MG
Encompass_Preferred
SGI-1776 free base is a novel ATP competitive inhibitor of Pim1 with IC50 of 7 nM in a cell-free assay, 50- and 10-fold selective versus Pim2 and Pim3, also potent to Flt3 and haspin. SGI-1776 induces apoptosis and autophagy. Phase 1. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-869-8