Learn More
Medchemexpress LLC SGK1-IN-3 hydrochloride | 96.7% | C23H21Cl3N6O3S | 100 MG
SDP
Supplier:  Medchemexpress LLC HY142686A100MG
Encompass_Preferred
SGK1-IN-3 hydrochloride is a potent SGK1 inhibitor with an IC50 of less than 1 μM. This compound shows potential for research in osteoarthritis.
  • Potent SGK1 inhibitor
  • IC50 of <1 μM
  • Potential for osteoarthritis research
  • Solid appearance
  • Light yellow to yellow color
  • Soluble in DMSO (125 mg/mL)
Catalog No. 50-003-53498