Learn More
Sino Biological Recombinant Human SGK1/SGK Protein (GST Tag), Active
Supplier:  Sino Biological S0611G500
Encompass_Preferred
Recombinant human SGK1 (60-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-257-3176