Learn More
Sino Biological Peptide Substrates: SGKtide
Supplier:  Sino Biological S0858
Encompass_Preferred
The SGKtide peptide sequence (CKKRNRRLSVA) contains serine protein kinase phosphorylation site and is evaluated as a substrate for SGK and NDR family kinases.
Catalog No. 50-256-7891