Learn More
Cayman Chemical SGlycylH1152hydrochLR 10mg
SDP
Supplier:  Cayman Chemical 1333210 MG
Encompass_Preferred
A selective and potent ROCK inhibitor (IC50 = 11.8 nM for ROCK2); poorly inhibits CaMKII, PKG, and Aurora A (IC50 = 2.57, 2.35, and 3.26 μM, respectively) as well as PKA or PKC (IC50 ≥10 μM for EAch)
Catalog No. 50-295-2567