Learn More
Selleck Chemical LLC SH-4-54
SDP
Supplier:  Selleck Chemical LLC S733710MM/1ML
Encompass_Preferred
SH-4-54 is a potent STAT inhibitor with KD of 300 nM and 464 nM for STAT3 and STAT5 respectively
Catalog No. 50-000-04309