Learn More
BIOPHARMA PEG SCIENTIFIC INC SH-PEG-SH 1G
Supplier:  BIOPHARMA PEG SCIENTIFIC INC HO00300320K
Encompass_Preferred
SH-PEG-SH | Purity: 95% | Mol Wt: 20K | 68865-60-1 | 1g
Catalog No. NC2202192