Learn More
Cayman Chemical SH1152 hydrochloride
SDP
Supplier:  Cayman Chemical 10007653500 UG
Encompass_Preferred
A potent, specific, ATP-competitive, and cell permeable inhibitor of ROCK (Ki = 1.6 nM); more potent inhibitor of ROCK than either Y-27632 (Ki = 140 nM) or HA-1077 (Ki = 330 nM); poorly inhibits PKA, PKC, and MLCK
Catalog No. 50-289-1181