Learn More
Medchemexpress LLC SH5-07 | 1456632-41-9 | >=98.0% | 625.61 | C29H28F5N3O5S | 100 MG

Supplier: Medchemexpress LLC HY100494100MG
SH5-07 is a hydroxamic acid-based small-molecule inhibitor of STAT3 for research applications. It inhibits STAT3 DNA-binding activity (IC50 3.9 μM in vitro), reduces expression of STAT3-regulated genes, and shows antitumor effects in cells with constitutively active STAT3. Supplied as a solid or as a 10 mM solution in DMSO, it is provided with standard analytical documentation and storage guidance.
- Hydroxamic acid-based STAT3 inhibitor with IC50 3.9 μM.
- Purity ≥98.0%.
- Available as solid and 10 mM DMSO solution.
- Includes datasheet, SDS, and COA for quality control.
- Recommended storage: powder and solvent conditions for stability.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.