Learn More
MSC Whitney Rand Heavy Duty Shelving, 14 Gauge, 5 Shelf, 1 ea
Supplier:  MSC 89841373
Encompass
This heavy-duty shelving unit provides a robust storage solution for industrial and laboratory environments. Constructed with 14-gauge steel, it is engineered for demanding applications that require high load-bearing capacity and long-term durability.
  • Features a 5-shelf configuration for organized storage
  • Constructed from 14-gauge steel for enhanced strength
  • Overall dimensions of 36 in. W x 24 in. D x 84 in. H
Catalog No. 50-005-11223