Learn More
Grainger Shaft Grounding Ring For 5/8 in Shaft
Supplier:  Grainger 14R028
Encompass_Preferred
The AEGIS Shaft Grounding Ring is a one-piece stainless steel ring that provides a path to ground for induced shaft voltages. It has a split-ring design that allows for easy installation without having to remove the coupled equipment. This grounding ring fits shafts from 1-3/4" to 7-15/16" in diameter and has a durable construction that provides long service life. It comes with mounting hardware for secure installation. The one-piece design eliminates the need for hinges or springs that can wear out over time. This shaft grounding ring helps protect bearings from fluting damage caused by stray shaft currents.
Catalog No. 50-274-8765