Learn More
Kuhner Shaker Inc SHAKER CABINET ISF4-X WITHOUT
Supplier:  Kuhner Shaker Inc SMX1600
Encompass
Shaker Cabinet ISF4-X without cooling..Temperature range: ambient + 10C to 80C..Control of 0.1C, Accuracy +/- 0.3C at 37C..100% stainless steel interior..External Dimensions: H1916 x W1494 x D960mm..Weight: 750kg..FIVE YEAR WARRANTY HARDWARE, ..1 YEAR ...
Catalog No. 50-257-0052
Add to Cart
Request a Quote