Learn More
Chemglass Life Sciences BenchWaver Rocker, Undulating, 3D Laboratory, 120V
SDP
Supplier:  Chemglass Life Sciences CLS4026010
Encompass
• Undulating, 3-Dimensional Motion• Adjustable Speed and Tilt Angle• Large, 13 Inch Platform• Ideal for Staining, Washing and General Mixing• 2 Year WarrantyThe BenchWaver™ combines the basic rocking motion of traditional platform rockers with the circular motion of an orbital shaker. This combination is ideal for generating the optimum 3-dimensional undulating motion commonly preferred for molecular biology applications. The large 13.5 x 13 inch platform is supplied complete with a non-slip, rubber mat. An optional stacking platform is available for users requiring additional mixing capacity. BenchWaver™ is a reliable, versatile instrument and can be used in refrigerated or incubated environments from 2 to 60°C. Tubes and plates sold separately.
Catalog No. 50-143-558