Learn More
Grainger Shaker Rocker 3 to 80 rpm 6 x13 x16
Supplier:  Grainger 14N171
Encompass_Preferred
The Benchmark Scientific 2D Rocker Rocker is a rocking platform shaker designed for mixing and agitation applications. It features a see-through acrylic platform that measures 8 x 8 inches and provides an oscillating rocking motion with adjustable speed up to 80 rocks per minute. This compact unit has non-slip rubber feet, requires minimal benchtop space, and operates quietly. It can handle load capacities up to 10 lbs and is ideal for gels, immunoblots, and general mixing tasks in life science and lab settings. The 2D Rocker Rocker requires 120V AC power.
Catalog No. 50-275-2037