Learn More
Selleck Chemical LLC Shanzhiside methyl ester S9307-1MG
SDP
Supplier:  Selleck Chemical LLC S93071MG
Encompass_Preferred
Shanzhiside methylester is a principle effective iridoid glycoside of L rotata and serves as a small molecule glucagon-like peptide-1 (GLP-1) receptor agonist
Catalog No. 50-000-25475